Learn More
Abnova™ Human GAS7 Full-length ORF (NP_958836.1, 1 a.a. - 416 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008522-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Growth arrest-specific 7 is expressed primarily in terminally differentiated brain cells and predominantly in mature cerebellar Purkinje neurons. GAS7 plays a putative role in neuronal development. Several transcript variants encoding proteins which vary in the N-terminus have been described. [provided by RefSeq]
Sequence: MKPGMVPPPPGEESQTVILPPGWQSYLSPQGRRYYVNTTTNETTWERPSSSPGIPASPGSHRSSLPPTVNGYHASGTPAHPPETAHMSVRKSTGDSQNLGSSSPSKKQSKENTITINCVTFPHPDTMPEQQLLKPTEWSYCDYFWADKKDPQGNGTVAGFELLLQKQLKGKQMQKEMSEFIRERIKIEEDYAKNLAKLSQNSLASQEEGSLGEAWAQVKKSLADEAEVHLKFSAKLHSEVEKPLMNFRENFKKDMKKCDHHIADLRKQLASRYASVEKARKALTERQRDLEMKTQQLEIKLSNKTEEDIKKARRKSTQAGDDLMRCVDLYNQAQSKWFEEMVTTTLELERLEVERVEMIRQHLCQYTQLRHETDMFNQSTVEPVDQLLRKVDPAKDRELWVREHKTGNIRPVDMEISpecifications
NP_958836.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
74.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0394/MGC1348/MLL/GAS7 | |
GAS7 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8522 | |
GAS7 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKPGMVPPPPGEESQTVILPPGWQSYLSPQGRRYYVNTTTNETTWERPSSSPGIPASPGSHRSSLPPTVNGYHASGTPAHPPETAHMSVRKSTGDSQNLGSSSPSKKQSKENTITINCVTFPHPDTMPEQQLLKPTEWSYCDYFWADKKDPQGNGTVAGFELLLQKQLKGKQMQKEMSEFIRERIKIEEDYAKNLAKLSQNSLASQEEGSLGEAWAQVKKSLADEAEVHLKFSAKLHSEVEKPLMNFRENFKKDMKKCDHHIADLRKQLASRYASVEKARKALTERQRDLEMKTQQLEIKLSNKTEEDIKKARRKSTQAGDDLMRCVDLYNQAQSKWFEEMVTTTLELERLEVERVEMIRQHLCQYTQLRHETDMFNQSTVEPVDQLLRKVDPAKDRELWVREHKTGNIRPVDMEI | |
RUO | |
GAS7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |