missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GALNT7 Partial ORF (NP_059119, 559 a.a. - 657 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00051809-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16107356

  • 349.00€ / 10µg

Estimated Shipment: 11-07-2023
to see stock.
Add to basket

Description

Description

This gene encodes GalNAc transferase 7, a member of the GalNAc-transferase family. The enzyme encoded by this gene controls the initiation step of mucin-type O-linked protein glycosylation and transfer of N-acetylgalactosamine to serine and threonine amino acid residues. This enzyme is a type II transmembrane protein and shares common sequence motifs with other family members. Unlike other family members, this enzyme shows exclusive specificity for partially GalNAc-glycosylated acceptor substrates and shows no activity with non-glycosylated peptides. This protein may function as a follow-up enzyme in the initiation step of O-glycosylation. [provided by RefSeq]

Sequence: LGPCHRMGGNQLFRINEANQLMQYDQCLTKGADGSKVMITHCNLNEFKEWQYFKNLHRFTHIPSGKCLDRSEVLHQVFISNCDSSKTTQKWEMNNIHSV
Specifications

Specifications

NP_059119
Liquid
51809
GALNT7 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GALNAC-T7/GalNAcT7
GALNT7
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LGPCHRMGGNQLFRINEANQLMQYDQCLTKGADGSKVMITHCNLNEFKEWQYFKNLHRFTHIPSGKCLDRSEVLHQVFISNCDSSKTTQKWEMNNIHSV
RUO
GALNT7
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers