missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human Gage7 Full-Length Orf (Abm85267.1, 1 A.A.-117 A.A.) Recombinant Protein With Gst Tag At N-Terminal.
Recombinant proteins expressed from in vitro wheat germ system. Such proteins mirror the conformation and folding of the native proteins in the eukaryotic biological system.
335.00€ - 508.00€
Specifications
Accession Number | ABM85267.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 2579 |
Name | GAGE7 (Human) Recombinant Protein (P02) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16105281
|
Abnova™
H00002579-P02.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
16115281
|
Abnova™
H00002579-P02.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
- Good Folding
- Excellent amino acids labeling
Immunogen Sequence: MSWRGRSTYYWPRPRRYVQPPEMIGPMRPEQFSDEVEPATPEEGEPATQRQDPAAAQEGEDEGASAGQGPKPEAHSQEQGHPQTGCECEDGPDGQEMDPPNPEEVKTPEEGEKQSQC
Specifications
ABM85267.1 | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH=8.0 in the elution buffer. | |
GAGE7 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GAGE-7 | |
GAGE7 | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
2579 | |
Glutathione Sepharose 4 Fast Flow | |
MSWRGRSTYYWPRPRRYVQPPEMIGPMRPEQFSDEVEPATPEEGEPATQRQDPAAAQEGEDEGASAGQGPKPEAHSQEQGHPQTGCECEDGPDGQEMDPPNPEEVKTPEEGEKQSQC | |
RUO | |
GAGE7 | |
GST |
For Research Use Only