missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FZD2 Partial ORF (NP_001457, 192 a.a. - 245 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002535-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16195211

  • 528.00€ / 25µg

Estimated Shipment: 07-11-2023
to see stock.
Add to basket

Description

Description

Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The expression of the FZD2 gene appears to be developmentally regulated, with high levels of expression in fetal kidney and lung and in adult colon and ovary. [provided by RefSeq]

Sequence: YATLEHPFHCPRVLKVPSYLSYKFLGERDCAAPCEPARPDGSMFFSQEETRFAR
Specifications

Specifications

NP_001457
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
31.68kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
YATLEHPFHCPRVLKVPSYLSYKFLGERDCAAPCEPARPDGSMFFSQEETRFAR
RUO
FZD2
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
2535
FZD2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FZD2
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers