missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FZD10 Partial ORF (NP_009128.1, 96 a.a. - 183 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00011211-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193896

  • 349.00€ / 10µg

Estimated Shipment: 03-11-2023
to see stock.
Add to basket

Description

Description

This gene is a member of the frizzled gene family. Members of this family encode 7-transmembrane domain proteins that are receptors for the Wingless type MMTV integration site family of signaling proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. Using array analysis, expression of this intronless gene is significantly up-regulated in two cases of primary colon cancer. [provided by RefSeq]

Sequence: TEQVSTPIPACRVMCEQARLKCSPIMEQFNFKWPDSLDCRKLPNKNDPNYLCMEAPNNGSDEPTRGSGLFPPLFRPQRPHSAQEHPLK
Specifications

Specifications

NP_009128.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.42kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TEQVSTPIPACRVMCEQARLKCSPIMEQFNFKWPDSLDCRKLPNKNDPNYLCMEAPNNGSDEPTRGSGLFPPLFRPQRPHSAQEHPLK
RUO
FZD10
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
11211
FZD10 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CD350/FZ-10/FzE7/hFz10
FZD10
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers