missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FUNDC2 Partial ORF (NP_076423.2, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00065991-Q04.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16095635

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

Sequence: METSAPRAGSQVVATTARHSAAYRADPLRVSSRDKLTEMAASSQGNFEGNFESLDLAEFAKKQPWWRKLFGQESGPSAEKYSVATQLFIGGVTGWCTGFI
Specifications

Specifications

NP_076423.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
METSAPRAGSQVVATTARHSAAYRADPLRVSSRDKLTEMAASSQGNFEGNFESLDLAEFAKKQPWWRKLFGQESGPSAEKYSVATQLFIGGVTGWCTGFI
RUO
FUNDC2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
65991
FUNDC2 (Human) Recombinant Protein (Q04)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DC44/FLJ33773/HCBP6/HCC3/MGC131676/MGC2495/PD03104
FUNDC2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers