missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FUK Partial ORF (NP_659496, 169 a.a. - 279 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00197258-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16102367

  • 508.00€ / 25µg
Estimated Shipment:
to see stock.
Add to basket

Description

Description

The protein encoded by this gene belongs to the GHMP (galacto-, homoserine, mevalonate and phosphomevalonate) kinase family and catalyzes the phosphorylation of L-fucose to form beta-L-fucose 1-phosphate. This enzyme catalyzes the first step in the utilization of free L-fucose in glycoprotein and glycolipid synthesis. L-fucose may be important in mediating a number of cell-cell interactions such as blood group antigen recognition, inflammation, and metastatis. While several transcript variants may exist for this gene, the full-length nature of only one has been described to date. [provided by RefSeq]

Sequence: RGARVIALPGSPAYAQNHGVYLTDPQGLVLDIYYQGTEAEIQRCVRPDGRVPLVSGVVFFSVETAERLLATHVSPPLDACTYLGLDSGARPVQLSLFFDILHCMAENVTRE
Specifications

Specifications

NP_659496
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.95kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RGARVIALPGSPAYAQNHGVYLTDPQGLVLDIYYQGTEAEIQRCVRPDGRVPLVSGVVFFSVETAERLLATHVSPPLDACTYLGLDSGARPVQLSLFFDILHCMAENVTRE
RUO
FUK
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
197258
FUK (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
1110046B12Rik/FLJ39408
FUK
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers