missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FOXR1 Partial ORF (NP_859072.1, 1 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00283150-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16192677

  • 349.00€ / 10µg

Estimated Shipment: 30-10-2023
to see stock.
Add to basket

Description

Description

This gene encodes a member of the forkhead box (FOX) family of transcription factors. FOX family members are monomeric, helix-turn-helix proteins with a core DNA-binding domain of approximately 110 aa. Many FOX transcription factors play roles in determining cell fates during early development. This forkhead box protein lacks the C-terminal basic region found in many other FOX family members. It is located within the 11q23.3 region which is commonly deleted in neuroblastomas. [provided by RefSeq]

Sequence: MGNELFLAFTTSHLPLAEQKLARYKLRIVKPPKLPLEKKPNPDKDGPDYEPNLWMWVNPNIVYPPGKLEVSGRRKREDLTSTLPSSQPPQKEEDASCSE
Specifications

Specifications

NP_859072.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MGNELFLAFTTSHLPLAEQKLARYKLRIVKPPKLPLEKKPNPDKDGPDYEPNLWMWVNPNIVYPPGKLEVSGRRKREDLTSTLPSSQPPQKEEDASCSE
RUO
FOXR1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
283150
FOXR1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DLNB13/FOXN5/MGC149486
FOXR1
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers