missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FOXI1 Full-length ORF (NP_658982.1, 1 a.a. - 283 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002299-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16157857

  • 335.00€ / 10µg
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it is possible that this gene plays an important role in the development of the cochlea and vestibulum, as well as embryogenesis. Mutations in this gene may be associated with the common cavity phenotype. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MSSFDLPAPSPPRCSPQFPSIGQEPPEMNLYYENFFHPQGVPSPQRPSFEGGGEYGATPNPYLWFNGPTMTPPPYLPGPNASPFLPQAYGVQRPLLPSVSGLGGSDLGWLPIPSQEELMKLVRPPYSYSALIAMAIHGAPDKRLTLSQIYQYVADNFPFYNKSKAGWQNSIRHNLSLNDCFKKVPRDEDDPAYVSGGSPTSHPLVTPGLSPEPSDKTGQNSLTFNSFSPLTNLSNHSGGGDWANPMPTNMLSYGGSVLSQFSPHFYNSVNTSGVLYPREGTEV
Specifications

Specifications

NP_658982.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
57.20kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FKH10/FKHL10/FREAC6/HFH3/MGC34197
FOXI1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
2299
FOXI1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSSFDLPAPSPPRCSPQFPSIGQEPPEMNLYYENFFHPQGVPSPQRPSFEGGGEYGATPNPYLWFNGPTMTPPPYLPGPNASPFLPQAYGVQRPLLPSVSGLGGSDLGWLPIPSQEELMKLVRPPYSYSALIAMAIHGAPDKRLTLSQIYQYVADNFPFYNKSKAGWQNSIRHNLSLNDCFKKVPRDEDDPAYVSGGSPTSHPLVTPGLSPEPSDKTGQNSLTFNSFSPLTNLSNHSGGGDWANPMPTNMLSYGGSVLSQFSPHFYNSVNTSGVLYPREGTEV
RUO
FOXI1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers