missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FOXC2 Partial ORF (NP_005242.1, 421 a.a. - 501 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002303-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16158204

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

This gene belongs to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of mesenchymal tissues. [provided by RefSeq]

Sequence: AASWYLNHSGDLNHLPGHTFAAQQQTFPNVREMFNSHRLGIENSTLGESQVSGNASCQLPYRSTPPLYRHAAPYSYDCTKY
Specifications

Specifications

NP_005242.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.65kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AASWYLNHSGDLNHLPGHTFAAQQQTFPNVREMFNSHRLGIENSTLGESQVSGNASCQLPYRSTPPLYRHAAPYSYDCTKY
RUO
FOXC2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
2303
FOXC2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FKHL14/LD/MFH-1/MFH1
FOXC2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers