Learn More
Abnova™ Human FOXC2 Full-length ORF (AAI11590.1, 1 a.a. - 501 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002303-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of mesenchymal tissues. [provided by RefSeq]
Sequence: MQARYSVSDPNALGVVPYLSEQNYYRAAGSYGGMASPMGVYSGHPEQYSAGMGRSYAPYHHHQPAAPKDLVKPPYSYIALITMAIQNAPEKKITLNGIYQFIMDRFPFYRENKQGWQNSIRHNLSLNECFVKVPRDDKKPGKGSYWTLDPDSYNMFENGSFLRRRRRFKKKDVSKEKEERAHLKEPPPAASKGAPATPHLADAPKEAEKKVVIKSEAASPALPVITKVETLSPESALQGSPRSAASTPAGSPDGSLPEHHAAAPNGLPGFSVENIMTLRTSPPGGELSPGAGRAGLVVPPLALPYAAAPPAAYGQPCAQGLEAGAAGGYQCSMRAMSLYTGAERPAHMCVPPALDEALSDHPSGPTSPLSALNLAAGQEGALAATGHHHQHHGHHHPQAPPPPPAPQPQPTPQPGAAAAQAASWYLNHSGDLNHLPGHTFAAQQQTFPNVREMFNSHRLGIENSTLGESQVSGNASCQLPYRSTPPLYRHAAPYSYDCTKYSpecifications
AAI11590.1 | |
Liquid | |
2303 | |
FOXC2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MQARYSVSDPNALGVVPYLSEQNYYRAAGSYGGMASPMGVYSGHPEQYSAGMGRSYAPYHHHQPAAPKDLVKPPYSYIALITMAIQNAPEKKITLNGIYQFIMDRFPFYRENKQGWQNSIRHNLSLNECFVKVPRDDKKPGKGSYWTLDPDSYNMFENGSFLRRRRRFKKKDVSKEKEERAHLKEPPPAASKGAPATPHLADAPKEAEKKVVIKSEAASPALPVITKVETLSPESALQGSPRSAASTPAGSPDGSLPEHHAAAPNGLPGFSVENIMTLRTSPPGGELSPGAGRAGLVVPPLALPYAAAPPAAYGQPCAQGLEAGAAGGYQCSMRAMSLYTGAERPAHMCVPPALDEALSDHPSGPTSPLSALNLAAGQEGALAATGHHHQHHGHHHPQAPPPPPAPQPQPTPQPGAAAAQAASWYLNHSGDLNHLPGHTFAAQQQTFPNVREMFNSHRLGIENSTLGESQVSGNASCQLPYRSTPPLYRHAAPYSYDCTKY | |
RUO | |
FOXC2 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
82.06kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FKHL14/LD/MFH-1/MFH1 | |
FOXC2 | |
Yes | |
wheat germ expression system |