missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FLJ46380 Full-length ORF (ENSP00000344316, 1 a.a. - 244 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00388591-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16100494

  • 528.00€ / 25µg

Estimated Shipment: 12-07-2023
to see stock.
Add to basket

Description

Description

Sequence: MSGAIFGPLEGPSSLDAPSIHPLVCPLCHVQYERPCLLDCFHDFCAGCLRGRATDGRLTCPLCQHQTVLKGPSGLPPVDRLLQFLVDSSGDGVEAVRCANCDLECSEQAGAAGRVGEEQRVPGCTVPNACTCTQHVFRGRPGSGFSSTSLGHLGPKCEPHYTGGETEVQNKGLEPVSRQWQRLRPFDLGRAHWSPIQGGVVDLHRRGSPVCRPGPTLKGLCYPSGIEAATAQGRWGQHAVPSGL
Specifications

Specifications

ENSP00000344316
Liquid
388591
FLJ46380 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSGAIFGPLEGPSSLDAPSIHPLVCPLCHVQYERPCLLDCFHDFCAGCLRGRATDGRLTCPLCQHQTVLKGPSGLPPVDRLLQFLVDSSGDGVEAVRCANCDLECSEQAGAAGRVGEEQRVPGCTVPNACTCTQHVFRGRPGSGFSSTSLGHLGPKCEPHYTGGETEVQNKGLEPVSRQWQRLRPFDLGRAHWSPIQGGVVDLHRRGSPVCRPGPTLKGLCYPSGIEAATAQGRWGQHAVPSGL
RUO
RNF207
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
52.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C1orf188/FLJ32096/FLJ46380/FLJ46593
RNF207
Yes
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers