missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FLJ46111 Full-length ORF (AAH93663.1, 1 a.a. - 197 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00283102-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16119823

  • 335.00€ / 10µg
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

Sequence: MNKVELESLLEGLTDEINFLRQLYEEEIWELQSQISDTSVVLSMDSSRSLDMDSIIPEVKAQYKEIANGSWAEAEGMHQIKYEELQTLPGKHRNDLRYAKMEISEINRNISRLQAQTEGLKGQRVSLEAAIADAEQYGELAVKDANTKLSSWRPPCSGPSKTWCSSCAMEHQELMNVKLALDIKIATYRKLLEGEES
Specifications

Specifications

AAH93663.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
48.8kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC138211
FLJ46111
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
283102
FLJ46111 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNKVELESLLEGLTDEINFLRQLYEEEIWELQSQISDTSVVLSMDSSRSLDMDSIIPEVKAQYKEIANGSWAEAEGMHQIKYEELQTLPGKHRNDLRYAKMEISEINRNISRLQAQTEGLKGQRVSLEAAIADAEQYGELAVKDANTKLSSWRPPCSGPSKTWCSSCAMEHQELMNVKLALDIKIATYRKLLEGEES
RUO
FLJ46111
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers