missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FKRP Partial ORF (NP_077277.1, 396 a.a. - 494 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00079147-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16119946

  • 335.00€ / 10µg
Estimated Shipment: 06-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a protein which is targeted to the medial Golgi apparatus and is necessary for posttranslational modification of dystroglycan. Mutations in this gene have been associated with congenital muscular dystrophy, mental retardation, and cerebellar cysts. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq]

Sequence: KAVEGDFFRVQYSESNHLHVDLWPFYPRNGVMTKDTWLDHRQDVEFPEHFLQPLVPLPFAGFVAQAPNNYRRFLELKFGPGVIENPQYPNPALLSLTGS
Specifications

Specifications

NP_077277.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KAVEGDFFRVQYSESNHLHVDLWPFYPRNGVMTKDTWLDHRQDVEFPEHFLQPLVPLPFAGFVAQAPNNYRRFLELKFGPGVIENPQYPNPALLSLTGS
RUO
FKRP
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
79147
FKRP (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ12576/LGMD2I/MDC1C/MGC2991
FKRP
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers