Learn More
Abnova™ Human FHL3 Partial ORF (NP_004459.2, 3 a.a. - 102 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002275-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
LIM proteins are defined by the possession of a highly conserved double zinc finger motif called the LIM domain.[supplied by OMIM]
Sequence: ESFDCAKCNESLYGRKYIQTDSGPYCVPCYDNTFANTCAECQQLIGHDSRELFYEDRHFHEGCFRCCRCQRSLADEPFTCQDSELLCNDCYCSAFSSQCSSpecifications
NP_004459.2 | |
Liquid | |
2275 | |
FHL3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC19547/MGC23614/MGC8696/SLIM2 | |
FHL3 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ESFDCAKCNESLYGRKYIQTDSGPYCVPCYDNTFANTCAECQQLIGHDSRELFYEDRHFHEGCFRCCRCQRSLADEPFTCQDSELLCNDCYCSAFSSQCS | |
RUO | |
FHL3 | |
Wheat Germ (in vitro) | |
GST |