Learn More
R&D Systems™ Human FGF basic/FGF2/bFGF Magnetic Luminex™ Performance Assay
Product Code. 18451990
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18451990 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18451990 Supplier R&D Systems™ Supplier No. LUHM233
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
- Magnetic format allows for easier wash steps - Undergoes similar validation testing as Quantikine™ ELISA Kits - Precise and Specific - Requires a small sample volume (<50μL) - Run your assay in just 3-5 hours - Mass-calibrated standards for consistent results with every new lot of material - Enough reagents for one 96-well plate
TRUSTED_SUSTAINABILITY
Specifications
Product Type Magnetic Bead-based Multiplex Assay for the Luminex™ MAGPIX™ Platform
Quantity 1 Kit
Species Human
Binding Target FGF basic/FGF2
Bead Type Magnetic
Assay Sensitivity 4.91pg/mL
Assay Range 24.00 - 5750pg/mL
Gene ID (Entrez) 2247
Gene Symbol FGF2
Sample Type Cell Culture Supernatant, Plasma (EDTA), Serum
Sample Volume Cell Culture Supernates (50μL),Serum (13μL),EDTA Plasma (13μL)
Synonym Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin
Kit Contents
  • Microparticle Concentrate
  • Biotin-Antibody Concentrate
Content And Storage Store the unopened product at 2° to 8°C. Do not use past expiration date.