Learn More
Abnova™ Human FCN2 Full-length ORF (NP_004099.2, 1 a.a. - 313 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002220-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The product of this gene belongs to the ficolin family of proteins. This family is characterized by the presence of a leader peptide, a short N-terminal segment, followed by a collagen-like region, and a C-terminal fibrinogen-like domain. This gene is predominantly expressed in the liver, and has been shown to have carbohydrate binding and opsonic activities. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: MELDRAVGVLGAATLLLSFLGMAWALQAADTCPEVKMVGLEGSDKLTILRGCPGLPGAPGPKGEAGTNGKRGERGPPGPPGKAGPPGPNGAPGEPQPCLTGPRTCKDLLDRGHFLSGWHTIYLPDCRPLTVLCDMDTDGGGWTVFQRRVDGSVDFYRDWATYKQGFGSRLGEFWLGNDNIHALTAQGTSELRVDLVDFEDNYQFAKYRSFKVADEAEKYNLVLGAFVEGSAGDSLTFHNNQSFSTKDQDNDLNTGNCAVMFQGAWWYKNCHVSNLNGRYLRGTHGSFANGINWKSGKGYNYSYKVSEMKVRPASpecifications
NP_004099.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
60.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EBP-37/FCNL/P35/ficolin-2 | |
FCN2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
2220 | |
FCN2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MELDRAVGVLGAATLLLSFLGMAWALQAADTCPEVKMVGLEGSDKLTILRGCPGLPGAPGPKGEAGTNGKRGERGPPGPPGKAGPPGPNGAPGEPQPCLTGPRTCKDLLDRGHFLSGWHTIYLPDCRPLTVLCDMDTDGGGWTVFQRRVDGSVDFYRDWATYKQGFGSRLGEFWLGNDNIHALTAQGTSELRVDLVDFEDNYQFAKYRSFKVADEAEKYNLVLGAFVEGSAGDSLTFHNNQSFSTKDQDNDLNTGNCAVMFQGAWWYKNCHVSNLNGRYLRGTHGSFANGINWKSGKGYNYSYKVSEMKVRPA | |
RUO | |
FCN2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |