Learn More
Abcam Human FCGR1A knockout THP-1 cell line
Product Code. 32154677 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1,000,000 Cells
2 x 1,000,000 Cells
Unit Size:
Each
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32154677 1,000,000 Cells Each
32142354 2 x 1,000,000 Cells Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 32154677 Supplier Abcam Supplier No. ab2758431000000Cells_
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
FCGR1A KO cell line available to order. KO validated by Next Generation Sequencing. Free of charge wild type control available. Knockout achieved by CRISPR/Cas9; X = 1 bp deletion, 2 bp deletion; Frameshift: 99.99%. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/via...
FCGR1A KO cell line available to order. KO validated by Next Generation Sequencing. Free of charge wild type control available. Knockout achieved by CRISPR/Cas9; X = 1 bp deletion, 2 bp deletion; Frameshift: 99.99%. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 1,000,000 Cells