missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FCER1G Full-length ORF (NP_004097.1, 1 a.a. - 86 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002207-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16107737

  • 528.00€ / 25µg

Estimated Shipment: 12-07-2023
to see stock.
Add to basket

Description

Description

The high affinity IgE receptor is a key molecule involved in allergic reactions. It is a tetramer composed of 1 alpha, 1 beta, and 2 gamma chains. The gamma chains are also subunits of other Fc receptors. [provided by RefSeq]

Sequence: MIPAVVLLLLLLVEQAAALGEPQLCYILDAILFLYGIVLTLLYCRLKIQVRKAAITSYEKSDGVYTGLSTRNQETYETLKHEKPPQ
Specifications

Specifications

NP_004097.1
Liquid
2207
FCER1G (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MIPAVVLLLLLLVEQAAALGEPQLCYILDAILFLYGIVLTLLYCRLKIQVRKAAITSYEKSDGVYTGLSTRNQETYETLKHEKPPQ
RUO
FCER1G
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FCRG
FCER1G
Yes
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers