Learn More
Abnova™ Human FBXO9 Full-length ORF (NP_258441.1, 1 a.a. - 437 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00026268-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. Alternative splicing of this gene generates at least 3 transcript variants diverging at the 5' terminus. [provided by RefSeq]
Sequence: MAEAEEDCHSDTVRADDDEENESPAETDLQAQLQMFRAQWMFELAPGVSSSNLENRPCRAARGSLQKTSADTKGKQEQAKEEKARELFLKAVEEEQNGALYEAIKFYRRAMQLVPDIEFKITYTRSPDGDGVGNSYIEDNDDDSKMADLLSYFQQQLTFQESVLKLCQPELESSQIHISVLPMEVLMYIFRWVVSSDLDLRSLEQLSLVCRGFYICARDPEIWRLACLKVWGRSCIKLVPYTSWREMFLERPRVRFDGVYISKTTYIRQGEQSLDGFYRAWHQVEYYRYIRFFPDGHVMMLTTPEEPQSIVPRLRTRNTRTDAILLGHYRLSQDTDNQTKVFAVITKKKEEKPLDYKYRYFRRVPVQEADQSFHVGLQLCSSGHQRFNKLIWIHHSCHITYKSTGETAVSAFEIDKMYTPLFFARVRSYTAFSERPLSpecifications
NP_258441.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
77.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434C0118/FBX9/KIAA0936/NY-REN-57/VCIA1/dJ341E18.2 | |
FBXO9 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
26268 | |
FBXO9 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAEAEEDCHSDTVRADDDEENESPAETDLQAQLQMFRAQWMFELAPGVSSSNLENRPCRAARGSLQKTSADTKGKQEQAKEEKARELFLKAVEEEQNGALYEAIKFYRRAMQLVPDIEFKITYTRSPDGDGVGNSYIEDNDDDSKMADLLSYFQQQLTFQESVLKLCQPELESSQIHISVLPMEVLMYIFRWVVSSDLDLRSLEQLSLVCRGFYICARDPEIWRLACLKVWGRSCIKLVPYTSWREMFLERPRVRFDGVYISKTTYIRQGEQSLDGFYRAWHQVEYYRYIRFFPDGHVMMLTTPEEPQSIVPRLRTRNTRTDAILLGHYRLSQDTDNQTKVFAVITKKKEEKPLDYKYRYFRRVPVQEADQSFHVGLQLCSSGHQRFNKLIWIHHSCHITYKSTGETAVSAFEIDKMYTPLFFARVRSYTAFSERPL | |
RUO | |
FBXO9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |