Learn More
Abnova™ Human FBXO42 Full-length ORF (AAH43410.1, 1 a.a. - 716 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00054455-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Members of the F-box protein family, such as FBXO42, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]
Sequence: MASSSDSEDDSFMAVDQEETVLEGTMDQDEEPHPVLEAEETRHNRSMSELPEEVLEYILSFLSPYQEHKTAALVCKQWYRLIKGVAHQCYHGFMKAVQEGNIQWESRTYPYPGTPITQRFSHSACYYDANQSMYVFGGCTQSSCNAAFNDLWRLDLNSKEWIRPLASGSYPSPKAGATLVVYKDLLVLFGGWTRPSPYPLHQPERFFDEIHTYSPSKNWWNCIVTTHGPPPMAGHSSCVIDDKMIVFGGSLGSRQMSNDVWVLDLEQWAWSKPNISGPSPHPRGGQSQIVIDDATILILGGCGGPNALFKDAWLLHMHSGPWAWQPLKVENEEHGAPELWCHPACRVGQCVVVFSQAPSGRAPLSPSLNSRPSPISATPPALVPETREYRSQSPVRSMDEAPCVNGRWGTLRPRAQRQTPSGSREGSLSPARGDGSPILNGGSLSPGTAAVGGSLDSPVQAISPSTPSAPEGYDLKIGLSLAPRRGSLPDQKDLRLGSIDLNWDLKPASSSNPMDGMDNRTVGGSMRHPPEQTNGVHTPPHVASALAGAVSPGALRRSLEAIKAMSSKGPSASAALSPPLGSSPGSPGSQSLSSGETVPIPRPGPAQGDGHSLPPIARRLGHHPPQSLNVGKPLYQSMNCKPMQMYVLDIKDTKEKGRVKWKVFNSSSVVGPPETSLHTVVQGRGELIIFGGLMDKKQNVKYYPKTNALYFVRAKRSpecifications
AAH43410.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
104.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Fbx42/KIAA1332 | |
FBXO42 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
54455 | |
FBXO42 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASSSDSEDDSFMAVDQEETVLEGTMDQDEEPHPVLEAEETRHNRSMSELPEEVLEYILSFLSPYQEHKTAALVCKQWYRLIKGVAHQCYHGFMKAVQEGNIQWESRTYPYPGTPITQRFSHSACYYDANQSMYVFGGCTQSSCNAAFNDLWRLDLNSKEWIRPLASGSYPSPKAGATLVVYKDLLVLFGGWTRPSPYPLHQPERFFDEIHTYSPSKNWWNCIVTTHGPPPMAGHSSCVIDDKMIVFGGSLGSRQMSNDVWVLDLEQWAWSKPNISGPSPHPRGGQSQIVIDDATILILGGCGGPNALFKDAWLLHMHSGPWAWQPLKVENEEHGAPELWCHPACRVGQCVVVFSQAPSGRAPLSPSLNSRPSPISATPPALVPETREYRSQSPVRSMDEAPCVNGRWGTLRPRAQRQTPSGSREGSLSPARGDGSPILNGGSLSPGTAAVGGSLDSPVQAISPSTPSAPEGYDLKIGLSLAPRRGSLPDQKDLRLGSIDLNWDLKPASSSNPMDGMDNRTVGGSMRHPPEQTNGVHTPPHVASALAGAVSPGALRRSLEAIKAMSSKGPSASAALSPPLGSSPGSPGSQSLSSGETVPIPRPGPAQGDGHSLPPIARRLGHHPPQSLNVGKPLYQSMNCKPMQMYVLDIKDTKEKGRVKWKVFNSSSVVGPPETSLHTVVQGRGELIIFGGLMDKKQNVKYYPKTNALYFVRAKR | |
RUO | |
FBXO42 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |