missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FBXO42 Full-length ORF (AAH43410.1, 1 a.a. - 716 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00054455-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16160533

  • 335.00€ / 10µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

Members of the F-box protein family, such as FBXO42, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]

Sequence: MASSSDSEDDSFMAVDQEETVLEGTMDQDEEPHPVLEAEETRHNRSMSELPEEVLEYILSFLSPYQEHKTAALVCKQWYRLIKGVAHQCYHGFMKAVQEGNIQWESRTYPYPGTPITQRFSHSACYYDANQSMYVFGGCTQSSCNAAFNDLWRLDLNSKEWIRPLASGSYPSPKAGATLVVYKDLLVLFGGWTRPSPYPLHQPERFFDEIHTYSPSKNWWNCIVTTHGPPPMAGHSSCVIDDKMIVFGGSLGSRQMSNDVWVLDLEQWAWSKPNISGPSPHPRGGQSQIVIDDATILILGGCGGPNALFKDAWLLHMHSGPWAWQPLKVENEEHGAPELWCHPACRVGQCVVVFSQAPSGRAPLSPSLNSRPSPISATPPALVPETREYRSQSPVRSMDEAPCVNGRWGTLRPRAQRQTPSGSREGSLSPARGDGSPILNGGSLSPGTAAVGGSLDSPVQAISPSTPSAPEGYDLKIGLSLAPRRGSLPDQKDLRLGSIDLNWDLKPASSSNPMDGMDNRTVGGSMRHPPEQTNGVHTPPHVASALAGAVSPGALRRSLEAIKAMSSKGPSASAALSPPLGSSPGSPGSQSLSSGETVPIPRPGPAQGDGHSLPPIARRLGHHPPQSLNVGKPLYQSMNCKPMQMYVLDIKDTKEKGRVKWKVFNSSSVVGPPETSLHTVVQGRGELIIFGGLMDKKQNVKYYPKTNALYFVRAKR
Specifications

Specifications

AAH43410.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
104.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Fbx42/KIAA1332
FBXO42
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
54455
FBXO42 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MASSSDSEDDSFMAVDQEETVLEGTMDQDEEPHPVLEAEETRHNRSMSELPEEVLEYILSFLSPYQEHKTAALVCKQWYRLIKGVAHQCYHGFMKAVQEGNIQWESRTYPYPGTPITQRFSHSACYYDANQSMYVFGGCTQSSCNAAFNDLWRLDLNSKEWIRPLASGSYPSPKAGATLVVYKDLLVLFGGWTRPSPYPLHQPERFFDEIHTYSPSKNWWNCIVTTHGPPPMAGHSSCVIDDKMIVFGGSLGSRQMSNDVWVLDLEQWAWSKPNISGPSPHPRGGQSQIVIDDATILILGGCGGPNALFKDAWLLHMHSGPWAWQPLKVENEEHGAPELWCHPACRVGQCVVVFSQAPSGRAPLSPSLNSRPSPISATPPALVPETREYRSQSPVRSMDEAPCVNGRWGTLRPRAQRQTPSGSREGSLSPARGDGSPILNGGSLSPGTAAVGGSLDSPVQAISPSTPSAPEGYDLKIGLSLAPRRGSLPDQKDLRLGSIDLNWDLKPASSSNPMDGMDNRTVGGSMRHPPEQTNGVHTPPHVASALAGAVSPGALRRSLEAIKAMSSKGPSASAALSPPLGSSPGSPGSQSLSSGETVPIPRPGPAQGDGHSLPPIARRLGHHPPQSLNVGKPLYQSMNCKPMQMYVLDIKDTKEKGRVKWKVFNSSSVVGPPETSLHTVVQGRGELIIFGGLMDKKQNVKYYPKTNALYFVRAKR
RUO
FBXO42
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers