missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FBXO31 Full-length ORF (AAH12748.1, 1 a.a. - 367 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00079791-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194473

  • 528.00€ / 25µg

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

Members of the F-box protein family, such as FBXO31, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]

Sequence: MYLPPHDPHVDDPMRFKPLFRIHLMERKAATVECMYGHKGPHHGHIQIVKKDEFSTKCNQTDHHRMSGGRQEEFRTWLREEWGRTLEDIFHEHMQELILMKFIYTSQYDNCLTYRRIYLPPSRPDDLIKPGLFKGTYGSHGLEIVMLSFHGRRARGTKITGDPNIPAGQQTVEIDLRHRIQLPDLENQRNFNELSRIVLEVRERVRQEQQEGGHEAGEGRGRQGPRESQPSPAQPRAEAPSKGPDGTPGEDGGEPGDAVAAAEQPAQCGQGQPFVLPVGVSSRNEDYPRTCRMCFYGTGLIAGHGFTSPERTPGVFILFDEDRFGFVWLELKSFSLYSRVQATFRNADAPSPQAFDEMLKNIQSLTS
Specifications

Specifications

AAH12748.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
68.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp434B027/DKFZp434J1815/FBX14/FBXO14/FLJ22477/Fbx31/MGC15419/MGC9527/pp2386
FBXO31
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
79791
FBXO31 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MYLPPHDPHVDDPMRFKPLFRIHLMERKAATVECMYGHKGPHHGHIQIVKKDEFSTKCNQTDHHRMSGGRQEEFRTWLREEWGRTLEDIFHEHMQELILMKFIYTSQYDNCLTYRRIYLPPSRPDDLIKPGLFKGTYGSHGLEIVMLSFHGRRARGTKITGDPNIPAGQQTVEIDLRHRIQLPDLENQRNFNELSRIVLEVRERVRQEQQEGGHEAGEGRGRQGPRESQPSPAQPRAEAPSKGPDGTPGEDGGEPGDAVAAAEQPAQCGQGQPFVLPVGVSSRNEDYPRTCRMCFYGTGLIAGHGFTSPERTPGVFILFDEDRFGFVWLELKSFSLYSRVQATFRNADAPSPQAFDEMLKNIQSLTS
RUO
FBXO31
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers