missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FBXO17 Partial ORF (NP_079183, 34 a.a. - 133 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00115290-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16121477

  • 335.00€ / 10µg
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class and it contains an F-box domain. Alternative splicing of this gene results in 2 transcript variants encoding different isoforms. [provided by RefSeq]

Sequence: PPRSLVTRCRPVCRAWRDIVDGPTVWLLQLARDRSAEGRALYAVAQRCLPSNEDKEEFPLCALARYCLRAPFGRNLIFNSCGEQGFRGWEVEHGGNGWAI
Specifications

Specifications

NP_079183
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
PPRSLVTRCRPVCRAWRDIVDGPTVWLLQLARDRSAEGRALYAVAQRCLPSNEDKEEFPLCALARYCLRAPFGRNLIFNSCGEQGFRGWEVEHGGNGWAI
RUO
FBXO17
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
115290
FBXO17 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FBG4/FBX26/FBXO26/FLJ11798/FLJ25205/Fbx17/MGC9379
FBXO17
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers