missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FBXL11 Partial ORF (NP_036440, 742 a.a. - 840 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00022992-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16106832

  • 349.00€ / 10µg

Estimated Shipment: 07-07-2023
to see stock.
Add to basket

Description

Description

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class and, in addition to an F-box, contains at least 6 highly degenerated leucine-rich repeats. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: GAGPSDHHSASRDERFKRRQLLRLQATERTMVREKENNPSGKKELSEVEKAKIRGSYLTVTLQRPTKELHGTSIVPKLQAITASSANLRHSPRVLVQHC
Specifications

Specifications

NP_036440
Liquid
22992
FBXL11 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CXXC8/DKFZp434M1735/FBL11/FBL7/FLJ00115/FLJ46431/JHDM1A/KDM2A/KIAA1004/LILINA
FBXL11
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GAGPSDHHSASRDERFKRRQLLRLQATERTMVREKENNPSGKKELSEVEKAKIRGSYLTVTLQRPTKELHGTSIVPKLQAITASSANLRHSPRVLVQHC
RUO
FBXL11
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers