missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FBLN2 Full-length ORF (NP_001989.2, 28 a.a. - 1184 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002199-P02.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194861

  • 528.00€ / 25µg

Estimated Shipment: 31-10-2023
to see stock.
Add to basket

Description

Description

This gene encodes an extracellular matrix protein, which belongs to the fibulin family. This protein binds various extracellular ligands and calcium. It may play a role during organ development, in particular, during the differentiation of heart, skeletal and neuronal structures. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: APRQDCTGVECPPLENCIEEALEPGACCATCVQQGCACEGYQYYDCLQGGFVRGRVPAGQSYFVDFGSTECSCPPGGGKISCQFMLCPELPPNCIEAVVVADSCPQCGQVGCVHAGHKYAAGHTVHLPPCRACHCPDAGGELICYQLPGCHGNFSDAEEGDPERHYEDPYSYDQEVAEVEAATALGGEVQAGAVQAGAGGPPAALGGGSQPLSTIQAPPWPAVLPRPTAAAALGPPAPVQAKARRVTEDSEEEEEEEEEREEMAVTEQLAAGGHRGLDGLPTTAPAGPSLPIQEERAEAGARAEAGARPEENLILDAQATSRSTGPEGVTHAPSLGKAALVPTQAVPGSPRDPVKPSPHNILSTSLPDAAWIPPTREVPRKPQVLPHSHVEEDTDPNSVHSIPRSSPEGSTKDLIETCCAAGQQWAIDNDECLEIPESGTEDNVCRTAQRHCCVSYLQEKSCMAGVLGAKEGETCGAEDNDSCGISLYKQCCDCCGLGLRVRAEGQSCESNPNLGYPCNHVMLSCCEGEEPLIVPEVRRPPEPAAAPRRVSEAEMAGREALSLGTEAELPNSLPGDDQDECLLLPGELCQHLCINTVGSYHCACFPGFSLQDDGRTCRPEGHPPQPEAPQEPALKSEFSQVASNTIPLPLPQPNTCKDNGPCKQVCSTVGGSAICSCFPGYAIMADGVSCEDINECVTDLHTCSRGEHCVNTLGSFHCYKALTCEPGYALKDGECEDVDECAMGTHTCQPGFLCQNTKGSFYCQARQRCMDGFLQDPEGNCVDINECTSLSEPCRPGFSCINTVGSYTCQRNPLICARGYHASDDGTKCVDVNECETGVHRCGEGQVCHNLPGSYRCDCKAGFQRDAFGRGCIDVNECWASPGRLCQHTCENTLGSYRCSCASGFLLAADGKRCEDVNECEAQRCSQECANIYGSYQCYCRQGYQLAEDGHTCTDIDECAQGAGILCTFRCLNVPGSYQCACPEQGYTMTANGRSCKDVDECALGTHNCSEAETCHNIQGSFRCLRFECPPNYVQVSKTKCERTTCHDFLECQNSPARITHYQLNFQTGLLVPAHIFRIGPAPAFTGDTIALNIIKGNEEGYFGTRRLNAYTGVVYLQRAVLEPRDFALDVEMKLWRQGSVTTFLAKMHIFFTTFAL
Specifications

Specifications

NP_001989.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
152.90kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FBLN2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
2199
FBLN2 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
APRQDCTGVECPPLENCIEEALEPGACCATCVQQGCACEGYQYYDCLQGGFVRGRVPAGQSYFVDFGSTECSCPPGGGKISCQFMLCPELPPNCIEAVVVADSCPQCGQVGCVHAGHKYAAGHTVHLPPCRACHCPDAGGELICYQLPGCHGNFSDAEEGDPERHYEDPYSYDQEVAEVEAATALGGEVQAGAVQAGAGGPPAALGGGSQPLSTIQAPPWPAVLPRPTAAAALGPPAPVQAKARRVTEDSEEEEEEEEEREEMAVTEQLAAGGHRGLDGLPTTAPAGPSLPIQEERAEAGARAEAGARPEENLILDAQATSRSTGPEGVTHAPSLGKAALVPTQAVPGSPRDPVKPSPHNILSTSLPDAAWIPPTREVPRKPQVLPHSHVEEDTDPNSVHSIPRSSPEGSTKDLIETCCAAGQQWAIDNDECLEIPESGTEDNVCRTAQRHCCVSYLQEKSCMAGVLGAKEGETCGAEDNDSCGISLYKQCCDCCGLGLRVRAEGQSCESNPNLGYPCNHVMLSCCEGEEPLIVPEVRRPPEPAAAPRRVSEAEMAGREALSLGTEAELPNSLPGDDQDECLLLPGELCQHLCINTVGSYHCACFPGFSLQDDGRTCRPEGHPPQPEAPQEPALKSEFSQVASNTIPLPLPQPNTCKDNGPCKQVCSTVGGSAICSCFPGYAIMADGVSCEDINECVTDLHTCSRGEHCVNTLGSFHCYKALTCEPGYALKDGECEDVDECAMGTHTCQPGFLCQNTKGSFYCQARQRCMDGFLQDPEGNCVDINECTSLSEPCRPGFSCINTVGSYTCQRNPLICARGYHASDDGTKCVDVNECETGVHRCGEGQVCHNLPGSYRCDCKAGFQRDAFGRGCIDVNECWASPGRLCQHTCENTLGSYRCSCASGFLLAADGKRCEDVNECEAQRCSQECANIYGSYQCYCRQGYQLAEDGHTCTDIDECAQGAGILCTFRCLNVPGSYQCACPEQGYTMTANGRSCKDVD
RUO
FBLN2
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers