missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human FABP2/I-FABP Quantikine ELISA Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
Pack of 96
Description
The Human FABP2/I-FABP Quantikine ELISA Kit quantifies human FABP2/I-FABP in cell culture supernates (50 ul), cell lysates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), urine (50 ul).
Specifications
Specifications
| Assay Range | 15.60 - 1000pg/mL |
| Assay Sensitivity | 6.21pg/mL |
| Detectable Analytes | FABP2/I-FABP |
| Assay | Solid Phase ELISA |
| Conjugate | HRP |
| Cross Reactivity | <0.5% cross-reactivity observed with available related molecules; <50% cross-species reactivity observed with species tested |
| Immunoassay Kit Format | 96-well Strip Plate |
| Product Type | ELISA Kit |
| Sample Type | Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Serum, Urine |
| Specificity | Natural and Recombinant Human FABP2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction