Learn More
R&D Systems™ Human FAAH (NP_001432.2) VersaClone cDNA
Specifications
Specifications
| Gene | FAAH |
| Accession Number | NP_001432.2 |
| Gene ID (Entrez) | 2166 |
| Gene Alias | Anandamide amidohydrolase 1, EC 3.5.1, EC 3.5.1.99, EC 3.5.1.n2, FAAH1, FAAH-1, fatty acid amide hydrolase, fatty-acid amide hydrolase 1, MGC102823, MGC138146, Oleamide hydrolase 1, PSAB |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Product Line | R&D Systems |
| Quantity | 10 μg |