missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human EXOSC9 Partial ORF (NP_005024, 124 a.a. - 220 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005393-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16103005

  • 528.00€ / 25µg

Estimated Shipment: 07-07-2023
to see stock.
Add to basket

Description

Description

Sequence: DPNEREERVMDGLLVIAMNKHREICTIQSSGGIMLLKDQVLRCSKIAGVKVAEITELILKALENDQKVRKEGGKFGFAESIANQRITAFKMEKAPID
Specifications

Specifications

NP_005024
Liquid
5393
EXOSC9 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PM/Scl-75/PMSCL1/RRP45/Rrp45p/p5/p6
EXOSC9
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.41kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
DPNEREERVMDGLLVIAMNKHREICTIQSSGGIMLLKDQVLRCSKIAGVKVAEITELILKALENDQKVRKEGGKFGFAESIANQRITAFKMEKAPID
RUO
EXOSC9
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers