missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human EXOSC10 Partial ORF (NP_001001998.1, 1 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00005394-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16123005

  • 359.00€ / 10µg

Estimated Shipment: 04-10-2022
to see stock.
Add to basket

Description

Description

Sequence: MAPPSTREPRVLSATSATKSDGEMVLPGFPDADSFVKFALGSVVAVTKASGGLPQFGDEYDFYRSFPGFQAFCETQGDRLLQCMSRVMQYHGCRSNIKD
Specifications

Specifications

NP_001001998.1
Liquid
5394
EXOSC10 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PM-Scl/PM/Scl-100/PMSCL/PMSCL2/RRP6/Rrp6p/p2/p3/p4
EXOSC10
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MAPPSTREPRVLSATSATKSDGEMVLPGFPDADSFVKFALGSVVAVTKASGGLPQFGDEYDFYRSFPGFQAFCETQGDRLLQCMSRVMQYHGCRSNIKD
RUO
EXOSC10
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers