Learn More
Abnova™ Human EXOC3 Full-length ORF (NP_009208.2, 1 a.a. - 745 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00011336-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. The complex is also essential for the biogenesis of epithelial cell surface polarity. [provided by RefSeq]
Sequence: MKETDREAVATAVQRVAGMLQRPDQLDKVEQYRRREARKKASVEARLKAAIQSQLDGVRTGLSQLHNALNDVKDIQQSLADVSKDWRQSINTIESLKDVKDAVVQHSQLAAAVENLKNIFSVPEIVRETQDLIEQGALLQAHRKLMDLECSRDGLMYEQYRMDSGNTRDMTLIHGYFGSTQGLSDELAKQLWMVLQRSLVTVRRDPTLLVSVVRIIEREEKIDRRILDRKKQTGFVPPGRPKNWKEKMFTILERTVTTRIEGTQADTRESDKMWLVRHLEIIRKYVLDDLIVAKNLMVQCFPPHYEIFKNLLNMYHQALSTRMQDLASEDLEANEIVSLLTWVLNTYTSTEMMRNVELAPEVDVGTLEPLLSPHVVSELLDTYMSTLTSNIIAWLRKALETDKKDWVKETEPEADQDGYYQTTLPAIVFQMFEQNLQVAAQISEDLKTKVLVLCLQQMNSFLSRYKDEAQLYKEEHLRNRQHPHCYVQYMIAIINNCQTFKESIVSLKRKYLKNEVEEGVSPSQPSMDGILDAIAKEGCSGLLEEVFLDLEQHLNELMTKKWLLGSNAVDIICVTVEDYFNDFAKIKKPYKKRMTAEAHRRVVVEYLRAVMQKRISFRSPEERKEGAEKMVREAEQLRFLFRKLASGFGEDVDGYCDTIVAVAEVIKLTDPSLLYLEVSTLVSKYPDIRDDHIGALLAVRGDASRDMKQTIMETLEQGPAQASPSYVPLFKDIVVPSLNVAKLLKSpecifications
NP_009208.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
112kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SEC6/SEC6L1/Sec6p | |
EXOC3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
11336 | |
EXOC3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKETDREAVATAVQRVAGMLQRPDQLDKVEQYRRREARKKASVEARLKAAIQSQLDGVRTGLSQLHNALNDVKDIQQSLADVSKDWRQSINTIESLKDVKDAVVQHSQLAAAVENLKNIFSVPEIVRETQDLIEQGALLQAHRKLMDLECSRDGLMYEQYRMDSGNTRDMTLIHGYFGSTQGLSDELAKQLWMVLQRSLVTVRRDPTLLVSVVRIIEREEKIDRRILDRKKQTGFVPPGRPKNWKEKMFTILERTVTTRIEGTQADTRESDKMWLVRHLEIIRKYVLDDLIVAKNLMVQCFPPHYEIFKNLLNMYHQALSTRMQDLASEDLEANEIVSLLTWVLNTYTSTEMMRNVELAPEVDVGTLEPLLSPHVVSELLDTYMSTLTSNIIAWLRKALETDKKDWVKETEPEADQDGYYQTTLPAIVFQMFEQNLQVAAQISEDLKTKVLVLCLQQMNSFLSRYKDEAQLYKEEHLRNRQHPHCYVQYMIAIINNCQTFKESIVSLKRKYLKNEVEEGVSPSQPSMDGILDAIAKEGCSGLLEEVFLDLEQHLNELMTKKWLLGSNAVDIICVTVEDYFNDFAKIKKPYKKRMTAEAHRRVVVEYLRAVMQKRISFRSPEERKEGAEKMVREAEQLRFLFRKLASGFGEDVDGYCDTIVAVAEVIKLTDPSLLYLEVSTLVSKYPDIRDDHIGALLAVRGDASRDMKQTIMETLEQGPAQASPSYVPLFKDIVVPSLNVAKLLK | |
RUO | |
EXOC3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |