Learn More
R&D Systems™ Human Erythropoietin/EPO DuoSet ELISA
Click to view available options
Quantity:
1 Kit (5 Plates)
Unit Size:
Pack of 5
Description
The Human Erythropoietin DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Erythropoietin.
Specifications
Specifications
| Assay Range | 3.12 - 200mIU/mL |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 2056 |
| Quantity | 1 Kit (5 Plates) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Synonym | ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2 |
| Target | Erythropoietin/EPO |