Learn More
Abnova™ Human ERP29 Full-length ORF (NP_006808.1, 1 a.a. - 261 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010961-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a reticuloplasmin, a protein which resides in the lumen of the endoplasmic reticulum (ER). The protein shows sequence similarity to the protein disulfide isomerase family. However, it lacks the thioredoxin motif characteristic of this family, suggesting that this protein does not function as a disulfide isomerase. The protein dimerizes and is thought to play a role in the processing of secretory proteins within the ER. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: MAAAVPRAAFLSPLLPLLLGFLLLSAPHGGSGLHTKGALPLDTVTFYKVIPKSKFVLVKFDTQYPYGEKQDEFKRLAENSASSDDLLVAEVGISDYGDKLNMELSEKYKLDKESYPVFYLFRDGDFENPVPYTGAVKVGAIQRWLKGQGVYLGMPGCLPVYDALAGEFIRASGVEARQALLKQGQDNLSSVKETQKKWAEQYLKIMGKILDQGEDFPASEMTRIARLIEKNKMSDGKKEELQKSLNILTAFQKKGAEKEELSpecifications
NP_006808.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
55.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C12orf8/ERp28/ERp31/PDI-DB | |
ERP29 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10961 | |
ERP29 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAAVPRAAFLSPLLPLLLGFLLLSAPHGGSGLHTKGALPLDTVTFYKVIPKSKFVLVKFDTQYPYGEKQDEFKRLAENSASSDDLLVAEVGISDYGDKLNMELSEKYKLDKESYPVFYLFRDGDFENPVPYTGAVKVGAIQRWLKGQGVYLGMPGCLPVYDALAGEFIRASGVEARQALLKQGQDNLSSVKETQKKWAEQYLKIMGKILDQGEDFPASEMTRIARLIEKNKMSDGKKEELQKSLNILTAFQKKGAEKEEL | |
RUO | |
ERP29 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |