missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ERCC1 Partial ORF (AAH52813, 207 a.a. - 281 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002067-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16107774

  • 528.00€ / 25µg

Estimated Shipment: 17-07-2023
to see stock.
Add to basket

Description

Description

Sequence: RYLETYKAYEQKPADLLMEKLEQDFVSRVTECLTTVKSVNKTDSQTLLTTFGSLEQLIAASREDLALCPGVGPQK
Specifications

Specifications

AAH52813
Liquid
2067
ERCC1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
COFS4/UV20
ERCC1
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.99kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RYLETYKAYEQKPADLLMEKLEQDFVSRVTECLTTVKSVNKTDSQTLLTTFGSLEQLIAASREDLALCPGVGPQK
RUO
ERCC1
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers