missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ERC1 Partial ORF (NP_055879.1, 850 a.a. - 947 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00023085-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16104416

  • 528.00€ / 25µg

Estimated Shipment: 06-07-2023
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is a member of a family of RIM-binding proteins. RIMs are active zone proteins that regulate neurotransmitter release. This gene has been found fused to the receptor-type tyrosine kinase gene RET by gene rearrangement due to the translocation t(10;12)(q11;p13). Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: KHLEEVLEMKQEALLAAISEKDANIALLELSSSKKKTQEEVAALKREKDRLVQQLKQQTQNRMKLMADNYEDDHFKSSHSNQTNHKPSPDQDEEEGIW
Specifications

Specifications

NP_055879.1
Liquid
23085
ERC1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Cast2/ELKS/KIAA1081/MGC12974/RAB6IP2
ERC1
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
KHLEEVLEMKQEALLAAISEKDANIALLELSSSKKKTQEEVAALKREKDRLVQQLKQQTQNRMKLMADNYEDDHFKSSHSNQTNHKPSPDQDEEEGIW
RUO
ERC1
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers