Learn More
Abnova™ Human EPM2AIP1 Full-length ORF (NP_055620.1, 1 a.a. - 607 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00009852-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The EPM2A gene, which encodes laforin, is mutated in an autosomal recessive form of adolescent progressive myoclonus epilepsy. The protein encoded by this gene binds to laforin, but its function is not known. This gene is intronless. [provided by RefSeq]
Sequence: MWMTPKRSKMEVDEALVFRPEWTQRYLVVEPPEGDGALCLVCRRLIVATRERDVRRHYEAEHEYYERYVADGERAALVERLRQGDLPVASFTPEERAARAGLGLCRLLALKGRGWGEGDFVYQCMEVLLREVLPEHVSVLQGVDLSPDITRQRILSIDRNLRNQLFNRARDFKAYSLALDDQAFVAYENYLLVFIRGVGPELEVQEDLLTIINLTHHFSVGALMSAILESLQTAGLSLQRMVGLTTTHTLRMIGENSGLVSYMREKAVSPNCWNVIHYSGFLHLELLSSYDVDVNQIINTISEWIVLIKTRGVRRPEFQTLLTESESEHGERVNGRCLNNWLRRGKTLKLIFSLRKEMEAFLVSVGATTVHFSDKQWLCDFGFLVDIMEHLRELSEELRVSKVFAAAAFDHICTFEVKLNLFQRHIEEKNLTDFPALREVVDELKQQNKEDEKIFDPDRYQMVICRLQKEFERHFKDLRFIKKDLELFSNPFNFKPEYAPISVRVELTKLQANTNLWNEYRIKDLGQFYAGLSAESYPIIKGVACKVASLFDSNQICEKAFSYLTRNQHTLSQPLTDEHLQALFRVATTEMEPGWDDLVRERNESNPSpecifications
NP_055620.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
96.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ11207/KIAA0766 | |
EPM2AIP1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
9852 | |
EPM2AIP1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MWMTPKRSKMEVDEALVFRPEWTQRYLVVEPPEGDGALCLVCRRLIVATRERDVRRHYEAEHEYYERYVADGERAALVERLRQGDLPVASFTPEERAARAGLGLCRLLALKGRGWGEGDFVYQCMEVLLREVLPEHVSVLQGVDLSPDITRQRILSIDRNLRNQLFNRARDFKAYSLALDDQAFVAYENYLLVFIRGVGPELEVQEDLLTIINLTHHFSVGALMSAILESLQTAGLSLQRMVGLTTTHTLRMIGENSGLVSYMREKAVSPNCWNVIHYSGFLHLELLSSYDVDVNQIINTISEWIVLIKTRGVRRPEFQTLLTESESEHGERVNGRCLNNWLRRGKTLKLIFSLRKEMEAFLVSVGATTVHFSDKQWLCDFGFLVDIMEHLRELSEELRVSKVFAAAAFDHICTFEVKLNLFQRHIEEKNLTDFPALREVVDELKQQNKEDEKIFDPDRYQMVICRLQKEFERHFKDLRFIKKDLELFSNPFNFKPEYAPISVRVELTKLQANTNLWNEYRIKDLGQFYAGLSAESYPIIKGVACKVASLFDSNQICEKAFSYLTRNQHTLSQPLTDEHLQALFRVATTEMEPGWDDLVRERNESNP | |
RUO | |
EPM2AIP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |