Learn More
Abcam Human EPCAM knockout A-431 cell line
Product Code. 32155763 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1,000,000 Cells
2 x 1,000,000 Cells
Unit Size:
Each
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32155763 2 x 1,000,000 Cells Pack of 2
32139339 1,000,000 Cells Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 32155763 Supplier Abcam Supplier No. ab2619022x1000000Cells_
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
EPCAM KO cell line available to order. KO validated by Immunocytochemistry, Next Generation Sequencing, Western blot. Free of charge wild type control available. Knockout achieved by CRISPR/Cas9 X = 2 bp deletion Frameshift: 99.58%. To order both knockout and wild-type control cells: select 2 x ...
EPCAM KO cell line available to order. KO validated by Immunocytochemistry, Next Generation Sequencing, Western blot. Free of charge wild type control available. Knockout achieved by CRISPR/Cas9 X = 2 bp deletion Frameshift: 99.58%. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 2 x 1,000,000 Cells