missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human EP400NL Full-length ORF (AAH66974.1, 1 a.a. - 419 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00347918-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MQHVSSSQSSQRHVQWPGACPGAGEEQPACSQPSLPLTLPSPSHQLQQLMVQTQSPTQPSPGPGQALQNVRAGAPGPGLGLCSSSPTGDFVDASVLVRQISLSPSSGGHFVFQDGSGLTQIAQGAQVQLQHPGTPITVRERRPSQPHTQSGGTIHHLGPQSPAAAGGAGLQPLASPSHITTANLPPQISSIIQGQLVQQQQVLQGPPLPRPLGFERTPGVLLPGAGGAAGFGMTSPPPPTSPSRTAVPPGLSSLPLTSVGNTGMKKVPKKLEEIPPASPEMAQMRKQCLDYHHQEMQALKEVFKEYLIELFFLQHFQGNMMDFLAFKERLYGPLQAYLRQNDLDIEEEEEEHFEVINDEVKVVARKHGQPGTPVAIATQLPPRTSAAFPAQQQPLQVLSDGSTVQLPRLSSLGFEDSMCSpecifications
AAH66974.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
71kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434I225/FLJ33915/FLJ44224/FLJ46340/MGC87589 | |
EP400NL | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
347918 | |
EP400NL (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MQHVSSSQSSQRHVQWPGACPGAGEEQPACSQPSLPLTLPSPSHQLQQLMVQTQSPTQPSPGPGQALQNVRAGAPGPGLGLCSSSPTGDFVDASVLVRQISLSPSSGGHFVFQDGSGLTQIAQGAQVQLQHPGTPITVRERRPSQPHTQSGGTIHHLGPQSPAAAGGAGLQPLASPSHITTANLPPQISSIIQGQLVQQQQVLQGPPLPRPLGFERTPGVLLPGAGGAAGFGMTSPPPPTSPSRTAVPPGLSSLPLTSVGNTGMKKVPKKLEEIPPASPEMAQMRKQCLDYHHQEMQALKEVFKEYLIELFFLQHFQGNMMDFLAFKERLYGPLQAYLRQNDLDIEEEEEEHFEVINDEVKVVARKHGQPGTPVAIATQLPPRTSAAFPAQQQPLQVLSDGSTVQLPRLSSLGFEDSMC | |
RUO | |
EP400NL | |
Wheat Germ (in vitro) | |
GST | |
Liquid |