Learn More
Human EMC2 (TTC35) knockout HeLa cell lysate, 1 Kit
Product Code. 32153694 Shop All Abcam Products
Product Code. 32153694 Supplier Abcam Supplier No. ab2631871Kit
 Restricted Item
Estimated Shipment: 10-11-2026
Human EMC2 (TTC35) knockout HeLa cell/tissue lysate.
  • Species: Human
  • Tissue: EMC2 (TTC35) knockout HeLa