Learn More
Abnova™ Human ELA1 Partial ORF (NP_001962.3, 159 a.a. - 257 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_001962.3 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1990 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16157554
|
Abnova™
H00001990-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
16147554
|
Abnova™
H00001990-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
Description
Elastases form a subfamily of serine proteases that hydrolyze many proteins in addition to elastin. Humans have six elastase genes which encode the structurally similar proteins elastase 1, 2, 2A, 2B, 3A, and 3B. Unlike other elastases, pancreatic elastase 1 is not expressed in the pancreas. To date, elastase 1 expression has only been detected in skin keratinocytes. Clinical literature that describes human elastase 1 activity in the pancreas or fecal material is actually referring to chymotrypsin-like elastase family, member 3B. [provided by RefSeq]
Sequence: LAQTLQQAYLPSVDYAICSSSSYWGSTVKNTMVCAGGDGVRSGCQGDSGGPLHCLVNGKYSVHGVTSFVSSRGCNVSRKPTVFTQVSAYISWINNVIASSpecifications
NP_001962.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ELA1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1990 | |
ELA1 (Human) Recombinant Protein (Q01) | |
LAQTLQQAYLPSVDYAICSSSSYWGSTVKNTMVCAGGDGVRSGCQGDSGGPLHCLVNGKYSVHGVTSFVSSRGCNVSRKPTVFTQVSAYISWINNVIAS | |
RUO | |
ELA1 | |
Recombinant | |
wheat germ expression system |