Learn More
Abcam Human EIF4H knockout HEK-293T cell line
Description
EIF4H KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: 1 bp insertion in exon 1. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Quantity | 2 x 1,000,000 Cells |
| Regulatory Status | RUO |