missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human EIF3EIP Full-length ORF (NP_057175.1, 1 a.a. - 564 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00051386-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16159972

  • 335.00€ / 10µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

Sequence: MSYPADDYESEAAYDPYAYPSDYDMHTGDPKQDLAYERQYEQQTYQVIPEVIKNFIQYFHKTVSDLIDQKVYELQASRVSSDVIDQKVYEIQDIYENSWTKLTERFFKNTPWPEAEAIAPQVGNDAVFLILYKELYYRHIYAKVSGGPSLEQRFESYYNYCNLFNYILNADGPAPLELPNQWLWDIIDEFIYQFQSFSQYRCKTAKKSEEEIDFLRSNPKIWNVHSVLNVLHSLVDKSNINRQLEVYTSGGDPESVAGEYGRHSLYKMLGYFSLVGLLRLHSLLGDYYQAIKVLENIELNKKSMYSRVPECQVTTYYYVGFAYLMMRRYQDAIRVFANILLYIQRTKSMFQRTTYKYEMINKQNEQMHALLAIALTMYPMRIDESIHLQLREKYGDKMLRMQKGDPQVYEELFSYSCPKFLSPVVPNYDNVHPNYHKEPFLQQLKVFSDEVQQQAQLSTIRSFLKLYTTMPVAKLAGFLDLTEQEFRIQLLVFKHKMKNLVWTSGISALDGEFQSASEVDFYIDKDMIHIADTKVARRYGDFFIRQIHKFEELNRTLKKMGQRP
Specifications

Specifications

NP_057175.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
93.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EIF3EIP/EIF3S11/EIF3S6IP/HSPC021/HSPC025/MSTP005
EIF3L
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
51386
EIF3EIP (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSYPADDYESEAAYDPYAYPSDYDMHTGDPKQDLAYERQYEQQTYQVIPEVIKNFIQYFHKTVSDLIDQKVYELQASRVSSDVIDQKVYEIQDIYENSWTKLTERFFKNTPWPEAEAIAPQVGNDAVFLILYKELYYRHIYAKVSGGPSLEQRFESYYNYCNLFNYILNADGPAPLELPNQWLWDIIDEFIYQFQSFSQYRCKTAKKSEEEIDFLRSNPKIWNVHSVLNVLHSLVDKSNINRQLEVYTSGGDPESVAGEYGRHSLYKMLGYFSLVGLLRLHSLLGDYYQAIKVLENIELNKKSMYSRVPECQVTTYYYVGFAYLMMRRYQDAIRVFANILLYIQRTKSMFQRTTYKYEMINKQNEQMHALLAIALTMYPMRIDESIHLQLREKYGDKMLRMQKGDPQVYEELFSYSCPKFLSPVVPNYDNVHPNYHKEPFLQQLKVFSDEVQQQAQLSTIRSFLKLYTTMPVAKLAGFLDLTEQEFRIQLLVFKHKMKNLVWTSGISALDGEFQSASEVDFYIDKDMIHIADTKVARRYGDFFIRQIHKFEELNRTLKKMGQRP
RUO
EIF3L
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers