missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human EFHD1 Full-length ORF (AAH04128.2, 1 a.a. - 239 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00080303-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16124913

  • 508.00€ / 25µg
Estimated Shipment: 27-05-2024
to see stock.
Add to basket

Description

Description

EFHD1 is an EF-hand domain-containing protein that displays increased expression during neuronal differentiation (Tominaga and Tomooka, 2002 [PubMed 12270117]).[supplied by OMIM]

Sequence: MASEELACKLERRLRREEAEESGPQLAPLGAPAPEPKPEPEPPARAPTASADAELSAQLSRRLDINEGAARPRRCRVFNPYTEFPEFSRRLIKDLESMFKLYDAGRDGFIDLMELKLMMEKLGAPQTHLGLKSMIKEVDEDFDGKLSFREFLLIFHKAAAGELQEDSGLMALAKLSEIDVALEGVRGAKNFFEAKVQALSSASKFEAELKAEQDERKREEEERRLRQAAFQKLKANFNT
Specifications

Specifications

AAH04128.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
53.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp781H0842/FLJ13612/MST133/MSTP133/PP3051
EFHD1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
80303
EFHD1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MASEELACKLERRLRREEAEESGPQLAPLGAPAPEPKPEPEPPARAPTASADAELSAQLSRRLDINEGAARPRRCRVFNPYTEFPEFSRRLIKDLESMFKLYDAGRDGFIDLMELKLMMEKLGAPQTHLGLKSMIKEVDEDFDGKLSFREFLLIFHKAAAGELQEDSGLMALAKLSEIDVALEGVRGAKNFFEAKVQALSSASKFEAELKAEQDERKREEEERRLRQAAFQKLKANFNT
RUO
EFHD1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers