Learn More
Abnova™ Human EEF1G Partial ORF (AAH15813.1, 33 a.a. - 89 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001937-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This subunit contains an N-terminal glutathione transferase domain, which may be involved in regulating the assembly of multisubunit complexes containing this elongation factor and aminoacyl-tRNA synthetases. [provided by RefSeq]
Sequence: SAPPHFHFGQTNRTPEFLRKFPAGKVPAFEGDDGFCVFESNAIAYYVSNEELRGSTPSpecifications
AAH15813.1 | |
Liquid | |
1937 | |
EEF1G (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EF1G/GIG35 | |
EEF1G | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.01kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SAPPHFHFGQTNRTPEFLRKFPAGKVPAFEGDDGFCVFESNAIAYYVSNEELRGSTP | |
RUO | |
EEF1G | |
Wheat Germ (in vitro) | |
GST |