missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human E2F7 Partial ORF (NP_976328.1, 801 a.a. - 910 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00144455-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16191997

  • 528.00€ / 25µg

Estimated Shipment: 30-10-2023
to see stock.
Add to basket

Description

Description

E2F transcription factors, such as E2F7, play an essential role in the regulation of cell cycle progression (Di Stefano et al., 2003 [PubMed 14633988]).[supplied by OMIM]

Sequence: VVNPKSSTLPSADPQLQSQPSLNLSPVMSRSHSVVQQPESPVYVGHPVSVVKLHQSPVPVTPKSIQRTHRETFFKTPGSLGDPVLKRRERNQSRNTSSAQRRLEIPSGGA
Specifications

Specifications

NP_976328.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VVNPKSSTLPSADPQLQSQPSLNLSPVMSRSHSVVQQPESPVYVGHPVSVVKLHQSPVPVTPKSIQRTHRETFFKTPGSLGDPVLKRRERNQSRNTSSAQRRLEIPSGGA
RUO
E2F7
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
144455
E2F7 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ12981
E2F7
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers