missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human E2F2 Partial ORF (NP_004082, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001870-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16194281

  • 528.00€ / 25µg

Estimated Shipment: 08-11-2023
to see stock.
Add to basket

Description

Description

The protein encoded by this gene is a member of the E2F family of transcription factors. The E2F family plays a crucial role in the control of cell cycle and action of tumor suppressor proteins and is also a target of the transforming proteins of small DNA tumor viruses. The E2F proteins contain several evolutionally conserved domains found in most members of the family. These domains include a DNA binding domain, a dimerization domain which determines interaction with the differentiation regulated transcription factor proteins (DP), a transactivation domain enriched in acidic amino acids, and a tumor suppressor protein association domain which is embedded within the transactivation domain. This protein and another 2 members, E2F1 and E2F3, have an additional cyclin binding domain. This protein binds specifically to retinoblastoma protein pRB in a cell-cycle dependent manner, and it exhibits overall 46% amino acid identity to E2F1. [provided by RefSeq]

Sequence: MLQGPRALASAAGQTPKVVPAMSPTELWPSGLSSPQLCPATATYYTPLYPQTAPPAAAPGTCLDATPHGPEGQVVRCLPAGRLPAKRKLDLEGIGRPVVPEFPTPKGKCI
Specifications

Specifications

NP_004082
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MLQGPRALASAAGQTPKVVPAMSPTELWPSGLSSPQLCPATATYYTPLYPQTAPPAAAPGTCLDATPHGPEGQVVRCLPAGRLPAKRKLDLEGIGRPVVPEFPTPKGKCI
RUO
E2F2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
1870
E2F2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
E2F-2
E2F2
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers