missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DYNC1I1 Full-length ORF (AAH53542.1, 1 a.a. - 628 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001780-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16117117

  • 508.00€ / 25µg
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

Sequence: MSDKSDLKAELERKKQRLAQIREEKKRKEEERKKKEADMQQKKEPVQDDSDLDRKRRETEALLQSIGISPEPPLVPTPMSPSSKSVSTPSEAGSQDSGDLGPLTRTLQWDTDPSVLQLQSDSELGRRLHKLGVSKVTQVDFLPREVVSYSKETQTPLATHQSEEDEEDEEMVESKVGQDSELENQDKKQEVKEAPPRELTEEEKQQILHLEEFLIFFDRTIRVIERALAEDSDIFFDYSGRELEEKDGDVQAGANLSFNRQFYDEHWSKHRVVTCMDWSLQYPELMVASYNNNEDAPHEPDGVALVWNMKFKKTTPEYVFHCQSSVMSVCFARFHPNLVVGGTYSGQIVLWDNRSHRRTPVQRTPLSAAAHTHPVYCVNVVGTQNAHNLITVSTDGKMCSWSLDMLSTPQESMELVYNKSKPVAVTGMAFPTGDVNNFVVGSEEGTVYTACRHGSKAGIGEVFEGHQGPVTGINCHMAVGPIDFSHLFVTSSFDWTVKLWTTKHNKPLYSFEDNADYVYDVMWSPVHPALFACVDGMGRLDLWNLNNDTEVPTASVAIEGASALNRVRWAQAGKEVAVGDSEGRIWVYDVGELAVPHNDEWTRFARTLVEIRANRADSEEEGTVELSA
Specifications

Specifications

AAH53542.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
97.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DNCI1/DNCIC1
DYNC1I1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
1780
DYNC1I1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSDKSDLKAELERKKQRLAQIREEKKRKEEERKKKEADMQQKKEPVQDDSDLDRKRRETEALLQSIGISPEPPLVPTPMSPSSKSVSTPSEAGSQDSGDLGPLTRTLQWDTDPSVLQLQSDSELGRRLHKLGVSKVTQVDFLPREVVSYSKETQTPLATHQSEEDEEDEEMVESKVGQDSELENQDKKQEVKEAPPRELTEEEKQQILHLEEFLIFFDRTIRVIERALAEDSDIFFDYSGRELEEKDGDVQAGANLSFNRQFYDEHWSKHRVVTCMDWSLQYPELMVASYNNNEDAPHEPDGVALVWNMKFKKTTPEYVFHCQSSVMSVCFARFHPNLVVGGTYSGQIVLWDNRSHRRTPVQRTPLSAAAHTHPVYCVNVVGTQNAHNLITVSTDGKMCSWSLDMLSTPQESMELVYNKSKPVAVTGMAFPTGDVNNFVVGSEEGTVYTACRHGSKAGIGEVFEGHQGPVTGINCHMAVGPIDFSHLFVTSSFDWTVKLWTTKHNKPLYSFEDNADYVYDVMWSPVHPALFACVDGMGRLDLWNLNNDTEVPTASVAIEGASALNRVRWAQAGKEVAVGDSEGRIWVYDVGELAVPHNDEWTRFARTLVEIRANRADSEEEGTVELSA
RUO
DYNC1I1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers