Learn More
Abcam Human DUSP4 knockout A549 cell line [F7]
Product Code. 32156125
Change view
Click to view available options
Quantity:
1,000,000 Cells
2 x 1,000,000 Cells
Unit Size:
Each
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32156125 1,000,000 Cells Each
32154082 2 x 1,000,000 Cells Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 32156125 Supplier Abcam Supplier No. ab3190901000000Cells_
 Restricted Item
Estimated Shipment: 20-10-2026
DUSP4 KO cell line available to order. KO validated by Next Generation Sequencing, Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9,109 bp deletion (allele 1) and 110 bp deletion (allele 2) in exon 2, CCDS6072.1. To order both knockout and wild-type...
DUSP4 KO cell line available to order. KO validated by Next Generation Sequencing, Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9,109 bp deletion (allele 1) and 110 bp deletion (allele 2) in exon 2, CCDS6072.1. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 1,000,000 Cells