Learn More
Abcam Human DUSP4 knockout A549 cell line [F7]
Description
DUSP4 KO cell line available to order. KO validated by Next Generation Sequencing, Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9,109 bp deletion (allele 1) and 110 bp deletion (allele 2) in exon 2, CCDS6072.1. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Quantity | 1,000,000 Cells |
| Regulatory Status | RUO |