Learn More
Abnova™ Human DUSP16 Partial ORF (NP_085143.1, 561 a.a. - 665 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00080824-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The activation of mitogen-activated protein kinase (MAPK) cascades transduces various extracellular signals to the nucleus to induce gene expression, cell proliferation, differentiation, cell cycle arrest, and apoptosis. For full activation of MAPKs, dual-specificity kinases phosphorylate both threonine and tyrosine residues in MAPK TXY motifs. MKPs are dual-specificity phosphatases that dephosphorylate the TXY motif, thereby negatively regulating MAPK activity.[supplied by OMIM]
Sequence: TESSHFYSASAIYGGSASYSAYSCSQLPTCGDQVYSVRRRQKPSDRADSRRSWHEESPFEKQFKRRSCQMEFGESIMSENRSREELGKVGSQSSFSGSMEIIEVSSpecifications
NP_085143.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.29kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TESSHFYSASAIYGGSASYSAYSCSQLPTCGDQVYSVRRRQKPSDRADSRRSWHEESPFEKQFKRRSCQMEFGESIMSENRSREELGKVGSQSSFSGSMEIIEVS | |
RUO | |
DUSP16 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
80824 | |
DUSP16 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA1700/MGC129701/MGC129702/MKP-7/MKP7 | |
DUSP16 | |
Recombinant | |
wheat germ expression system |