Learn More
Abnova™ Human DSC2 Partial ORF (NP_077740.1, 444 a.a. - 534 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001824-Q02.10ug
Additional Details : Weight : 0.02000kg
Description
The protein encoded by this gene is a calcium-dependent glycoprotein that is a member of the desmocollin subfamily of the cadherin superfamily. These desmosomal family members, along with the desmogleins, are found primarily in epithelial cells where they constitute the adhesive proteins of the desmosome cell-cell junction and are required for cell adhesion and desmosome formation. The desmosomal family members are arranged in two clusters on chromosome 18, occupying less than 650 kb combined. Mutations in this gene are associated with arrhythmogenic right ventricular dysplasia-11. Alternative splicing results in two transcript variants encoding distinct isoforms. [provided by RefSeq]
Sequence: SREASPRSAMSTATVTVNVEDQDEGPECNPPIQTVRMKENAEVGTTSNGYKAYDPETRSSSGIRYKKLTDPTGWVTIDENTGSIKVFRSLDSpecifications
NP_077740.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.75kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SREASPRSAMSTATVTVNVEDQDEGPECNPPIQTVRMKENAEVGTTSNGYKAYDPETRSSSGIRYKKLTDPTGWVTIDENTGSIKVFRSLD | |
RUO | |
DSC2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1824 | |
DSC2 (Human) Recombinant Protein (Q02) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARVD11/CDHF2/DG2/DGII/III/DKFZp686I11137/DSC3 | |
DSC2 | |
Recombinant | |
wheat germ expression system |